Assay Details
Target Gene Details
Entrez Gene ID: | 170779 |
Gene Name: | CD209d antigen |
Gene Aliases: |
SIGN-R3, SIGNR3, mSIGNR3 |
Location: |
Chr.8:3871824-3878548 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd209d | NM_130904.2 | NP_570974.1 | ||
| AF373411.1 | AAL13237.1 | |||
| AF440280.1 | AAL33584.1 | |||
| AK136586.1 | BAE23062.1 |
More Information
Set Membership: |
|

Genomic Map