Assay Details
Target Gene Details
Entrez Gene ID: | 234374 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 49 |
Gene Aliases: |
R27090_2 |
Location: |
Chr.8:70292866-70302511 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx49 | NM_001024922.2 | NP_001020093.2 | ||
| AK139073.1 | BAE23880.1 | |||
| BC099554.1 | AAH99554.1 |

Genomic Map