Assay Details
Target Gene Details
Entrez Gene ID: | 654824 |
Gene Name: | ankyrin repeat domain 37 |
Gene Aliases: |
- |
Location: |
Chr.8:45996908-46000237 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd37 | NM_001039562.1 | NP_001034651.1 | ||
| AK020869.1 | ||||
| AK075868.1 | ||||
| BC039642.1 | ||||
| BC092377.1 | AAH92377.1 |

Genomic Map