Assay Details
Target Gene Details
Entrez Gene ID: | 353188 |
Gene Name: | a disintegrin and metallopeptidase domain 32 |
Gene Aliases: |
- |
Location: |
Chr.8:24836143-24948917 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adam32 | NM_001293693.1 | NP_001280622.1 | ||
| NM_153397.2 | NP_700446.2 | |||
| AF513715.1 | AAN03858.1 | |||
| AK005759.1 | BAC25124.1 | |||
| AK131749.1 | ||||
| BC060983.1 | AAH60983.1 |
More Information
Set Membership: |
|

Genomic Map