Assay Details
Target Gene Details
Entrez Gene ID: | 234564 |
Gene Name: | carboxylesterase 1F |
Gene Aliases: |
AU018778, CesML1, TGH-2 |
Location: |
Chr.8:93256236-93279766 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1f | NM_144930.2 | 12 | 1409 | NP_659179.1 |
| AB023632.1 | 8 | 812 | BAB60699.1 | |
| AI174063.1 | 3 | 517 | ||
| AK136491.1 | 12 | 1401 | BAE23005.1 | |
| BC013479.1 | 12 | 1384 | AAH13479.1 |
More Information
Set Membership: |
|

Genomic Map