Assay Details
Target Gene Details
Entrez Gene ID: | 234564 |
Gene Name: | carboxylesterase 1F |
Gene Aliases: |
AU018778, CesML1, TGH-2 |
Location: |
Chr.8:93256236-93279766 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1f | NM_144930.2 | NP_659179.1 | ||
| AK136491.1 | BAE23005.1 | |||
| BC013479.1 | AAH13479.1 |
More Information
Set Membership: |
|

Genomic Map