Assay Details
Target Gene Details
Entrez Gene ID: | 212392 |
Gene Name: | coiled-coil domain containing 110 |
Gene Aliases: |
Gm172 |
Location: |
Chr.8:45934649-45945230 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccdc110 | NM_001033246.2 | NP_001028418.1 | ||
| AK132742.1 | BAE21328.1 | |||
| AK161248.1 | BAE36267.1 |
More Information
Set Membership: |
|

Genomic Map