Assay Details
Target Gene Details
Entrez Gene ID: | 12623 |
Gene Name: | carboxylesterase 1G |
Gene Aliases: |
Ces-1, Ces1, Ses-1 |
Location: |
Chr.8:93302369-93337209 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1g | NM_021456.4 | NP_067431.2 | ||
| AK136446.1 | BAE22982.1 | |||
| AK136588.1 | BAE23063.1 | |||
| AK149446.1 | BAE28882.1 | |||
| AK162328.1 | BAE36856.1 | |||
| BC021150.1 | AAH21150.1 | |||
| BC026897.1 | AAH26897.1 | |||
| Y12887.1 | CAA73388.1 |
More Information
Set Membership: |
|

Genomic Map