Assay Details
Target Gene Details
Entrez Gene ID: | 102247 |
Gene Name: | 1-acylglycerol-3-phosphate O-acyltransferase 6 (lysophosphatidic acid acyltransferase, zeta) |
Gene Aliases: |
AU041707, AW545732, GPAT4, Tsarg7 |
Location: |
Chr.8:23171265-23209171 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agpat6 | NM_018743.4 | NP_061213.2 | ||
| AF406611.1 | AAP97283.1 | |||
| AK045235.1 | BAC32273.1 | |||
| AK049675.1 | ||||
| AK077561.1 | BAC36864.1 | |||
| AK083589.1 | BAC38962.1 | |||
| AK161270.1 | BAE36282.1 | |||
| AK167876.1 | BAE39890.1 | |||
| AK169257.1 | BAE41020.1 | |||
| AK178773.1 | ||||
| AK182757.1 | ||||
| AK188390.1 | ||||
| AK204634.1 | ||||
| AY489184.1 | AAS75838.1 | |||
| BC031767.1 | AAH31767.1 |
More Information
Set Membership: |
|

Genomic Map