Assay Details
Target Gene Details
Entrez Gene ID: | 434350 |
Gene Name: | RIKEN cDNA 9430091E24 gene |
Gene Aliases: |
- |
Location: |
Chr.8:111127444-111145498 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9430091E24Rik | NR_040363.1 | 4 | 468 | |
| NR_040364.1 | 3 | 345 | ||
| AK020517.1 | 4 | 452 | ||
| AK053982.1 | 3 | 307 | ||
| BF472197.1 | 3 | 344 |
Target Gene Details
Entrez Gene ID: | 319518 |
Gene Name: | pyruvate dehydrogenase phosphatase regulatory subunit |
Gene Aliases: |
4930402E16Rik, AU018056, mKIAA1990 |
Location: |
Chr.8:111094599-111139779 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 16 - Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pdpr | NM_198308.1 | NP_938050.1 | ||
| AK029575.1 | ||||
| AK078515.1 | ||||
| AK129472.1 | BAC98282.1 | |||
| AY223867.1 | AAP20649.1 | |||
| BC120745.1 | ||||
| BC125425.1 |
More Information
Set Membership: |
|

Genomic Map