Assay Details
Target Gene Details
Entrez Gene ID: | 66929 |
Gene Name: | anti-silencing function 1B histone chaperone |
Gene Aliases: |
1700003K02Rik, AA409591 |
Location: |
Chr.8:83955694-83970195 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Asf1b | NM_024184.2 | 3 | 408 | NP_077146.1 |
| AK005646.1 | 3 | 441 | BAB24166.1 | |
| AK019594.1 | 3 | 190 | BAB31809.1 | |
| AK077005.1 | 3 | 434 | BAC36561.1 | |
| AK077718.1 | 3 | 389 | BAC36978.1 | |
| AK134906.1 | 3 | 596 | BAE22334.1 | |
| AK150818.1 | 3 | 389 | BAE29881.1 | |
| AK151951.1 | 3 | 389 | BAE30824.1 | |
| AK165322.1 | 3 | 387 | BAE38137.1 | |
| BC003428.1 | 3 | 408 | AAH03428.1 |
More Information
Set Membership: |
|

Genomic Map