Assay Details
Target Gene Details
Entrez Gene ID: | 22428 |
Gene Name: | dynactin 6 |
Gene Aliases: |
AU044699, WS-3, p27 |
Location: |
Chr.8:34090420-34108712 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dctn6 | NM_001293757.1 | NP_001280686.1 | ||
| NM_001293759.1 | NP_001280688.1 | |||
| NM_011722.4 | NP_035852.1 | |||
| AF124788.1 | AAD30477.1 | |||
| AF190796.1 | AAF05616.1 | |||
| AK003618.1 | BAB22892.1 | |||
| AK088312.1 | BAC40276.1 | |||
| BC029249.1 | AAH29249.1 | |||
| BY079663.1 | ||||
| CA573939.1 | ||||
| DV062065.1 |
Target Gene Details
Entrez Gene ID: | 234155 |
Gene Name: | membrane bound O-acyltransferase domain containing 4 |
Gene Aliases: |
GOAT, Gm171 |
Location: |
Chr.8:34108219-34125185 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mboat4 |
More Information
Set Membership: |
|

Genomic Map