Assay Details
Target Gene Details
Entrez Gene ID: | 11733 |
Gene Name: | ankyrin 1, erythroid |
Gene Aliases: |
Ank-1, nb, pale |
Location: |
Chr.8:22974836-23150502 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ank1 | NM_001110783.3 | NP_001104253.1 | ||
| NM_001277280.2 | NP_001264209.1 | |||
| NM_001277281.2 | NP_001264210.1 | |||
| NM_001277284.2 | NP_001264213.1 | |||
| NM_001277286.2 | NP_001264215.1 | |||
| NM_001277289.2 | NP_001264218.1 | |||
| NM_031158.4 | NP_112435.2 | |||
| NR_102385.2 | ||||
| NR_102386.2 | ||||
| AK147278.1 | BAE27815.1 | |||
| AK147483.1 | ||||
| AK147597.1 | BAE28015.1 | |||
| AK158131.1 | BAE34375.1 | |||
| BC079910.1 | ||||
| BC138029.1 | ||||
| BC171944.1 | ||||
| M84756.1 | AAA37236.1 | |||
| X69063.1 | CAA48801.1 | |||
| X69065.1 | CAA48803.1 |

Genomic Map