Assay Details
Target Gene Details
Entrez Gene ID: | 234138 |
Gene Name: | TELO2 interacting protein 2 |
Gene Aliases: |
- |
Location: |
Chr.8:31150316-31164703 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tti2 | NM_001199988.1 | 5 | 1766 | NP_001186917.1 |
| NM_144927.3 | 5 | 1766 | NP_659176.2 | |
| NR_103719.1 | 5 | 1766 | ||
| AK045392.1 | 5 | 1771 | BAC32341.1 | |
| AK046031.1 | 5 | 1756 | BAC32576.1 | |
| AK084242.1 | 5 | 1756 | BAC39146.1 | |
| AK139425.1 | 5 | 1743 | BAE24007.1 | |
| BC019943.1 | 5 | 1469 | AAH19943.1 |

Genomic Map