Assay Details
Target Gene Details
Entrez Gene ID: | 407789 |
Gene Name: | cDNA sequence BC048644 |
Gene Aliases: |
- |
Location: |
Chr.8:121907833-121918428 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC048644 | NM_001033485.2 | 2 | 1946 | NP_001028657.1 |
| AK133128.1 | 2 | 1947 | BAE21521.1 |
Target Gene Details
Entrez Gene ID: | 12352 |
Gene Name: | carbonic anhydrase 5a, mitochondrial |
Gene Aliases: |
CAV, Ca5a, Car5 |
Location: |
Chr.8:121916135-121944912 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car5a | NM_007608.2 | NP_031634.2 | ||
| AI098626.1 | ||||
| AI132225.1 | ||||
| AI314492.1 | ||||
| AI527458.1 | ||||
| BC030174.1 | AAH30174.1 | |||
| X51971.1 | CAA36233.1 |

Genomic Map