Assay Details
Target Gene Details
Entrez Gene ID: | 12354 |
Gene Name: | carbonic anhydrase 7 |
Gene Aliases: |
AV343731, Ca7 |
Location: |
Chr.8:104534700-104550347 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car7 | NM_001301164.1 | NP_001288093.1 | ||
| NM_053070.3 | NP_444300.1 | |||
| AK090390.1 | ||||
| AY075021.1 | AAL78169.1 | |||
| BC094913.1 | AAH94913.1 | |||
| BY741075.1 |

Genomic Map