Assay Details
Target Gene Details
Entrez Gene ID: | 12354 |
Gene Name: | carbonic anhydrase 7 |
Gene Aliases: |
AV343731, Ca7 |
Location: |
Chr.8:104534700-104550347 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Car7 | NM_001301164.1 | 2 | 582 | NP_001288093.1 |
| NM_001301165.1 | 2 | 150 | NP_001288094.1 | |
| NM_053070.3 | 2 | 177 | NP_444300.1 | |
| AF291660.1 | 1 | 72 | AAG16230.1 | |
| AK090390.1 | 2 | 180 | ||
| AK144275.1 | 2 | 151 | BAE25808.1 | |
| AY075021.1 | 2 | 178 | AAL78169.1 | |
| BC094913.1 | 2 | 165 | AAH94913.1 | |
| BY741075.1 | 2 | 582 |

Genomic Map