Assay Details
Target Gene Details
Entrez Gene ID: | 16598 |
Gene Name: | Kruppel-like factor 2 (lung) |
Gene Aliases: |
Lklf |
Location: |
Chr.8:72319062-72321654 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Klf2 | NM_008452.2 | NP_032478.2 | ||
| AK131962.1 | BAE20905.1 | |||
| BC138737.1 | ||||
| BC138738.1 | ||||
| BF023555.1 | ||||
| U25096.1 | AAA86728.1 |
More Information
Set Membership: |
|

Genomic Map