Assay Details
Target Gene Details
Entrez Gene ID: | 67619 |
Gene Name: | NIN1/RPN12 binding protein 1 homolog (S. cerevisiae) |
Gene Aliases: |
1700021I09Rik, ART-4, Nob1p, Psmd8bp1 |
Location: |
Chr.8:107412489-107425038 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nob1 | NM_026277.3 | NP_080553.1 | ||
| AK021194.1 | BAB32325.1 | |||
| AK054034.1 | BAC35627.1 | |||
| AK075755.1 | BAC35933.1 | |||
| AK165521.1 | ||||
| BC054835.1 | AAH54835.1 | |||
| BC103793.1 | AAI03794.1 | |||
| BC117992.2 | ||||
| BC117993.2 |

Genomic Map