Assay Details
Target Gene Details
Entrez Gene ID: | 71780 |
Gene Name: | myo-inositol 1-phosphate synthase A1 |
Gene Aliases: |
1300017C10Rik, AU018670 |
Location: |
Chr.8:70594481-70597290 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Isyna1 | NM_023627.1 | 7 | 897 | NP_076116.1 |
| AF288525.1 | 7 | 897 | AAF90201.1 | |
| AK005029.2 | 7 | 875 | BAB23756.1 | |
| AK079323.1 | 7 | 874 | BAC37607.1 | |
| AK137089.1 | 6 | 979 | ||
| BC003458.1 | 7 | 888 | AAH03458.1 |
More Information
Set Membership: |
|

Genomic Map