Assay Details
Target Gene Details
Entrez Gene ID: | 68133 |
Gene Name: | glycine cleavage system protein H (aminomethyl carrier) |
Gene Aliases: |
1100001L02Rik, 5730591C18Rik |
Location: |
Chr.8:116981828-116993449 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gcsh | NM_026572.3 | NP_080848.1 | ||
| AK003788.2 | BAB22996.2 | |||
| AK019983.1 | BAB31951.1 | |||
| AK050373.1 | BAC34217.1 | |||
| BC014745.1 | AAH14745.1 |

Genomic Map