Assay Details
Target Gene Details
Entrez Gene ID: | 244595 |
Gene Name: | carboxylesterase 1A |
Gene Aliases: |
Ces1l, EG244595, Gm4976 |
Location: |
Chr.8:93020214-93048192 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ces1a | NM_001013764.2 | NP_001013786.2 | ||
| BC089371.1 | AAH89371.1 |
More Information
Set Membership: |
|

Genomic Map