Assay Details
Target Gene Details
Entrez Gene ID: | 17259 |
Gene Name: | myocyte enhancer factor 2B |
Gene Aliases: |
AI451606 |
Location: |
Chr.8:70152778-70167488 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mef2b | NM_001045484.1 | NP_001038949.1 | ||
| NM_008578.2 | NP_032604.2 | |||
| BC045147.1 | AAH45147.1 | |||
| D87833.1 | BAA24542.1 | |||
| D87834.1 | BAA24543.1 | |||
| D87835.1 | BAA24544.1 | |||
| D87836.1 | BAA24545.1 |
More Information
Set Membership: |
|

Genomic Map