Assay Details
Target Gene Details
Entrez Gene ID: | 67184 |
Gene Name: | NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13 |
Gene Aliases: |
2700054G14Rik, AU022060, CDA016, CGI-39, GRIM-19, Grim19 |
Location: |
Chr.8:69887788-69902558 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufa13 | NM_023312.2 | NP_075801.1 | ||
| BC034899.1 | AAH34899.1 |
More Information
Set Membership: |
|

Genomic Map