Assay Details
Target Gene Details
Entrez Gene ID: | 26558 |
Gene Name: | homer homolog 3 (Drosophila) |
Gene Aliases: |
AW146114 |
Location: |
Chr.8:70282891-70294361 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Homer3 | NM_001146153.1 | NP_001139625.1 | ||
| NM_011984.2 | NP_036114.2 | |||
| AF093261.1 | AAC71025.1 | |||
| AK155463.1 | 1 | 3049 | ||
| AK179223.1 | ||||
| BC005773.1 | AAH05773.1 | |||
| BC095961.1 | AAH95961.1 | |||
| BU054773.1 | ||||
| CN539202.1 | ||||
| CX208892.1 |
More Information
Set Membership: |
|

Genomic Map