Assay Details
Target Gene Details
Entrez Gene ID: | 16816 |
Gene Name: | lecithin cholesterol acyltransferase |
Gene Aliases: |
AI046659, D8Wsu61e |
Location: |
Chr.8:105939551-105943402 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lcat | NM_008490.2 | NP_032516.2 | ||
| AI574112.1 | ||||
| AK004914.1 | BAB23665.1 | |||
| AK149476.1 | BAE28903.1 | |||
| BC028861.1 | AAH28861.1 | |||
| J05154.1 | AAA39419.1 |
More Information
Set Membership: |
|

Genomic Map