Assay Details
Target Gene Details
Entrez Gene ID: | 77411 |
Gene Name: | epithelial splicing regulatory protein 2 |
Gene Aliases: |
9530027K23Rik, Rbm35b |
Location: |
Chr.8:106130403-106137528 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esrp2 | NM_176838.2 | 14 | 2229 | NP_789808.1 |
| AK020578.1 | 1 | 57 | BAC25635.1 | |
| AK088755.1 | 14 | 2099 | ||
| AK168488.1 | 13 | 1462 | BAE40375.1 | |
| BC031444.1 | 14 | 2229 | AAH31444.1 |
More Information
Set Membership: |
|

Genomic Map