Assay Details
Target Gene Details
Entrez Gene ID: | 234155 |
Gene Name: | membrane bound O-acyltransferase domain containing 4 |
Gene Aliases: |
GOAT, Gm171 |
Location: |
Chr.8:34108219-34125185 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mboat4 | NM_001126314.2 | 1 | 123 | NP_001119786.1 |
| BC147844.1 | 1 | 109 | ||
| BC147849.1 | 1 | 108 | ||
| EU518496.1 | 1 | 59 | ||
| EU721729.1 | 1 | 123 |
More Information
Set Membership: |
|

Genomic Map