Assay Details
Target Gene Details
Entrez Gene ID: | 234384 |
Gene Name: | MPV17 mitochondrial membrane protein-like 2 |
Gene Aliases: |
Fksg24 |
Location: |
Chr.8:70758649-70760921 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mpv17l2 | NM_183170.2 | 4 | 518 | NP_898993.1 |
| AK138901.1 | 4 | 544 | BAE23813.1 | |
| AK151477.1 | 4 | 541 | BAE30432.1 | |
| AK152031.1 | 4 | 541 | BAE30890.1 | |
| AK168278.1 | 4 | 496 | BAE40224.1 | |
| AK188164.1 | 2 | 95 | ||
| AY013699.1 | 4 | 518 | AAG38937.1 | |
| BC051227.1 | 4 | 527 | AAH51227.1 |
More Information
Set Membership: |
|

Genomic Map