Assay Details
Target Gene Details
Entrez Gene ID: | 56749 |
Gene Name: | dihydroorotate dehydrogenase |
Gene Aliases: |
2810417D19Rik, AI834883 |
Location: |
Chr.8:109593244-109608701 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhodh | NM_020046.3 | 7 | 1240 | NP_064430.1 |
| AF029667.2 | 7 | 945 | AAB82948.2 | |
| AK013102.1 | ||||
| BC019542.1 | 7 | 950 | AAH19542.1 | |
| BC027829.1 | 7 | 1201 | AAH27829.1 | |
| BC045206.1 | 7 | 1190 | AAH45206.1 |
More Information
Set Membership: |
|

Genomic Map