Assay Details
Target Gene Details
Entrez Gene ID: | 17751 |
Gene Name: | metallothionein 3 |
Gene Aliases: |
Mt-3 |
Location: |
Chr.8:94152607-94154148 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mt3 | NM_013603.2 | NP_038631.1 | ||
| AK140209.1 | BAE24282.1 | |||
| BC059725.1 | AAH59725.1 |
More Information
Set Membership: |
|

Genomic Map