Assay Details
Target Gene Details
Entrez Gene ID: | 14128 |
Gene Name: | Fc receptor, IgE, low affinity II, alpha polypeptide |
Gene Aliases: |
CD23, Fce2, Fcer2, Ly-42 |
Location: |
Chr.8:3680726-3694174 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcer2a | NM_001253737.1 | NP_001240666.1 | ||
| NM_001253739.1 | NP_001240668.1 | |||
| NM_001253743.1 | NP_001240672.1 | |||
| NM_001253745.1 | NP_001240674.1 | |||
| NM_001253746.1 | NP_001240675.1 | |||
| NM_001253747.1 | NP_001240676.1 | |||
| NM_013517.4 | NP_038545.1 | |||
| AK156654.1 | BAE33796.1 | |||
| AK172591.1 | BAE43084.1 | |||
| AY069980.1 | AAL58516.1 | |||
| AY069981.1 | AAL58517.1 | |||
| BC119164.1 | ||||
| BC119190.1 | ||||
| M34163.1 | AAA37603.1 | |||
| M99371.1 | AAA74898.1 | |||
| S67072.1 | AAB28791.2 | |||
| S67073.1 | AAB28792.2 | |||
| S67074.1 | AAB28793.2 | |||
| X64223.1 | CAA45532.1 | |||
| X64224.1 | CAA45533.1 |
More Information
Set Membership: |
|

Genomic Map