Assay Details
Target Gene Details
Entrez Gene ID: | 16336 |
Gene Name: | insulin-like 3 |
Gene Aliases: |
Rlf, Rlnl |
Location: |
Chr.8:71689252-71690577 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Insl3 | NM_013564.7 | 1 | 89 | NP_038592.3 |
| AK132857.1 | 1 | 90 | BAE21395.1 | |
| BC049540.1 | 1 | 89 | AAH49540.1 | |
| S82815.1 | 1 | 88 | AAB39365.1 |
Target Gene Details
Entrez Gene ID: | 16453 |
Gene Name: | Janus kinase 3 |
Gene Aliases: |
fae, wil |
Location: |
Chr.8:71676383-71690577 on Build GRCm38 |
Assay Gene Location: | Within Intron 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Jak3 | NM_010589.6 | NP_034719.2 | ||
| AK038268.1 | BAC29956.1 | |||
| AK088365.1 | BAC40305.1 | |||
| BC105577.1 | ||||
| BC131646.1 | ||||
| BC131647.1 | ||||
| L32955.1 | AAA21565.1 | |||
| L33768.1 | AAA21415.1 |
More Information
Set Membership: |
|

Genomic Map