Assay Details
Target Gene Details
Entrez Gene ID: | 75809 |
Gene Name: | RIKEN cDNA 4930488L21 gene |
Gene Aliases: |
4930452J22Rik |
Location: |
Chr.8:93811694-93813555 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4930488L21Rik | NR_026888.1 | |||
| AK015446.1 | ||||
| AK015647.1 |
Target Gene Details
Entrez Gene ID: | 14681 |
Gene Name: | guanine nucleotide binding protein, alpha O |
Gene Aliases: |
AW050213, Galphao, Gnao, alphaO |
Location: |
Chr.8:93810561-93969388 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gnao1 | NM_001113384.1 | NP_001106855.1 | ||
| NM_010308.3 | NP_034438.1 | |||
| AK035907.1 | BAC29238.1 | |||
| AK047197.1 | BAC32987.1 | |||
| AK160434.1 | ||||
| BC051989.1 | AAH51989.1 | |||
| M36777.1 | AAA37645.1 | |||
| M36778.1 | AAA74566.1 |
More Information
Set Membership: |
|

Genomic Map