Assay Details
Target Gene Details
| Entrez Gene ID: | 14182 | 
| Gene Name: | fibroblast growth factor receptor 1 | 
| Gene Aliases: | AW208770, Eask, FLG, Fgfr-1, Flt-2, Hspy, MFR, bFGF-R-1, c-fgr | 
| Location: | Chr.8:25513623-25575718 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 11 - Exon 12 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Fgfr1 | NM_001079908.2 | NP_001073377.1 | ||
| NM_001079909.2 | NP_001073378.1 | |||
| NM_010206.3 | NP_034336.2 | |||
| AF176552.1 | AAF05312.1 | |||
| AK028354.1 | BAC25899.1 | |||
| AK034295.1 | BAC28662.1 | |||
| AK049704.1 | ||||
| AK167639.1 | BAE39690.1 | |||
| BC010200.1 | AAH10200.1 | |||
| BC033447.1 | AAH33447.1 | |||
| M28998.1 | AAA37290.1 | |||
| M33760.1 | AAA37622.1 | |||
| M65053.1 | AAA37620.1 | |||
| U22324.1 | AAC52182.1 | |||
| U23445.1 | AAC52183.1 | |||
| X51893.1 | CAA36175.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top