Assay Details
Target Gene Details
Entrez Gene ID: | 14182 |
Gene Name: | fibroblast growth factor receptor 1 |
Gene Aliases: |
AW208770, Eask, FLG, Fgfr-1, Flt-2, Hspy, MFR, bFGF-R-1, c-fgr |
Location: |
Chr.8:25513623-25575718 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fgfr1 | NM_001079908.2 | NP_001073377.1 | ||
| NM_001079909.2 | NP_001073378.1 | |||
| NM_010206.3 | NP_034336.2 | |||
| AF176552.1 | AAF05312.1 | |||
| AK028354.1 | BAC25899.1 | |||
| AK049704.1 | ||||
| BC010200.1 | AAH10200.1 | |||
| BU611199.1 | ||||
| CB524826.1 | ||||
| CF532036.1 | ||||
| M28998.1 | AAA37290.1 | |||
| M33760.1 | AAA37622.1 | |||
| S47233.1 | ||||
| S74765.1 | AAB32844.2 | |||
| U22324.1 | AAC52182.1 | |||
| U23445.1 | AAC52183.1 | |||
| X51893.1 | CAA36175.1 |
More Information
Set Membership: |
|

Genomic Map