Assay Details
Target Gene Details
Entrez Gene ID: | 94187 |
Gene Name: | zinc finger protein 423 |
Gene Aliases: |
Ebfaz, Roaz, Zfp104, Znf423, ataxia1, mKIAA0760, nur12 |
Location: |
Chr.8:87661810-87961859 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp423 | NM_033327.2 | NP_201584.2 | ||
| AF188609.1 | AAG17053.1 | |||
| AK122365.1 | BAC65647.1 | |||
| AK133497.1 | BAE21688.1 | |||
| AY147407.1 | AAN39840.1 | |||
| AY256893.1 | AAP33073.1 | |||
| BC059234.1 | AAH59234.1 | |||
| BC139028.1 | ||||
| BC139030.1 | ||||
| BU519630.1 |
More Information
Set Membership: |
|

Genomic Map