Assay Details
Target Gene Details
Entrez Gene ID: | 22702 |
Gene Name: | zinc finger protein 42 |
Gene Aliases: |
Rex-1, Rex1, Zfp-42 |
Location: |
Chr.8:43295067-43307009 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp42 | NM_009556.3 | NP_033582.2 | ||
| AK141232.1 | BAE24604.1 | |||
| BC053399.1 | AAH53399.1 | |||
| BC098467.1 | AAH98467.1 | |||
| M28382.1 | AAA40053.1 |
More Information
Set Membership: |
|

Genomic Map