Assay Details
Target Gene Details
Entrez Gene ID: | 57264 |
Gene Name: | resistin |
Gene Aliases: |
ADSF, Fizz3, Rstn, Xcp4 |
Location: |
Chr.8:3655770-3659818 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Retn | NM_001204959.1 | NP_001191888.1 | ||
| NM_022984.4 | NP_075360.1 | |||
| AF290870.1 | AAK83102.1 | |||
| AF323080.1 | AAG59823.1 | |||
| AK131613.1 | BAE20720.1 | |||
| AK131869.1 | BAE20842.1 | |||
| BC051196.1 | AAH51196.1 |

Genomic Map