Assay Details
Target Gene Details
Entrez Gene ID: | 20845 |
Gene Name: | steroidogenic acute regulatory protein |
Gene Aliases: |
AV363654, D8Ertd419e |
Location: |
Chr.8:25808513-25815982 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Star | NM_011485.4 | 1 | 60 | NP_035615.2 |
| AK014431.1 | 1 | 70 | ||
| AK019725.2 | 1 | 100 | BAB31842.1 | |
| AK054470.1 | 1 | 60 | BAC35791.1 | |
| AK136327.1 | 1 | 70 | BAE22934.1 | |
| BC060970.1 | 1 | 71 | AAH60970.1 | |
| BC082283.1 | 1 | 71 | AAH82283.1 | |
| L36062.1 | 1 | 76 | AAB94783.1 |

Genomic Map