Assay Details
Target Gene Details
Entrez Gene ID: | 78323 |
Gene Name: | family with sequence similarity 219, member B |
Gene Aliases: |
2310046O06Rik, AI594866 |
Location: |
Chr.9:57537528-57543188 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam219b | NM_001166364.1 | NP_001159836.1 | ||
| NM_175273.4 | NP_780482.1 | |||
| AK028365.1 | BAC25905.1 | |||
| AK185896.1 | ||||
| BC111883.1 | ||||
| BC112403.1 |

Genomic Map