Assay Details
Target Gene Details
Entrez Gene ID: | 22186 |
Gene Name: | ubiquitin A-52 residue ribosomal protein fusion product 1 |
Gene Aliases: |
D8Ertd21e, Gm1863, Rps27a, Ubb, Ubc |
Location: |
Chr.8:70508266-70510367 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uba52 | NM_019883.3 | NP_063936.1 | ||
| AF118402.1 | AAD14688.1 | |||
| AK018726.1 | BAB31371.1 | |||
| BC014772.1 | AAH14772.1 | |||
| BC054413.1 | AAH54413.1 | |||
| BC080838.1 | AAH80838.1 | |||
| BC086924.1 | AAH86924.1 | |||
| BC087922.1 | AAH87922.1 | |||
| EF437347.1 |

Genomic Map