Assay Details
Target Gene Details
Entrez Gene ID: | 382035 |
Gene Name: | poly(A)binding protein nuclear 1-like |
Gene Aliases: |
Gm1108, Pabpnl1, ePABP2 |
Location: |
Chr.8:122619471-122622735 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pabpn1l | NM_001007462.1 | NP_001007463.1 | ||
| BC141316.1 | ||||
| BC141317.1 | ||||
| BC145634.1 |
More Information
Set Membership: |
|

Genomic Map