Assay Details
Target Gene Details
Entrez Gene ID: | 192654 |
Gene Name: | phospholipase A2, group XV |
Gene Aliases: |
ACS, C87498, LLPL, Lpla2, Lypla3 |
Location: |
Chr.8:106150351-106164715 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g15 | NM_133792.2 | 1 | 103 | NP_598553.1 |
| AF468958.1 | AAL78651.1 | |||
| AK028614.1 | 1 | 103 | ||
| AK085194.1 | 1 | 92 | BAC39387.1 | |
| AK155004.1 | 1 | 99 | BAE32987.1 | |
| AK163111.1 | 1 | 92 | BAE37197.1 | |
| AK170814.1 | 1 | 91 | BAE42046.1 | |
| AK193243.1 | ||||
| AK215799.1 | 1 | 44 | ||
| BC019373.1 | 1 | 103 | AAH19373.1 |
More Information
Set Membership: |
|

Genomic Map