Assay Details
Target Gene Details
Entrez Gene ID: | 192654 |
Gene Name: | phospholipase A2, group XV |
Gene Aliases: |
ACS, C87498, LLPL, Lpla2, Lypla3 |
Location: |
Chr.8:106150351-106164715 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pla2g15 | NM_133792.2 | NP_598553.1 | ||
| AF468958.1 | AAL78651.1 | |||
| AK028614.1 | ||||
| AK085194.1 | BAC39387.1 | |||
| AK155004.1 | BAE32987.1 | |||
| AK163111.1 | BAE37197.1 | |||
| AK170814.1 | BAE42046.1 | |||
| AK177399.1 | ||||
| AK180645.1 | ||||
| AK186155.1 | ||||
| AK188333.1 | ||||
| AK189283.1 | ||||
| AK203519.1 | ||||
| AK205252.1 | ||||
| AK205480.1 | ||||
| AK207442.1 | ||||
| AK211077.1 | ||||
| AK212406.1 | ||||
| AK212507.1 | ||||
| AK212587.1 | ||||
| AK212623.1 | ||||
| AK214459.1 | ||||
| AK215050.1 | ||||
| AK215274.1 | ||||
| AK215692.1 | ||||
| AK215942.1 | ||||
| AK217196.1 | ||||
| AK219754.1 | ||||
| AK220071.1 | ||||
| BC019373.1 | AAH19373.1 |
More Information
Set Membership: |
|

Genomic Map