Assay Details
Target Gene Details
Entrez Gene ID: | 75692 |
Gene Name: | nuclear receptor 2C2-associated protein |
Gene Aliases: |
2310073E15Rik |
Location: |
Chr.8:70131327-70133752 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr2c2ap | NM_001025586.2 | NP_001020757.1 | ||
| NM_001025587.2 | NP_001020758.1 | |||
| NR_104434.1 | ||||
| AK010140.1 | ||||
| AK010864.1 | ||||
| AK160346.1 | BAE35750.1 | |||
| AK168268.1 | ||||
| BC092288.1 | AAH92288.1 | |||
| BY722132.1 | ||||
| CX222571.1 |
Target Gene Details
Entrez Gene ID: | 19727 |
Gene Name: | regulatory factor X-associated ankyrin-containing protein |
Gene Aliases: |
Tvl1 |
Location: |
Chr.8:70130806-70139197 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfxank | NM_001025589.1 | 9 | 1840 | NP_001020760.1 |
| NM_011266.2 | 9 | 1870 | NP_035396.1 | |
| AF094761.1 | AAC69884.1 | |||
| AK052298.1 | 9 | 1797 | BAC34924.1 | |
| AK087462.1 | 9 | 1771 | ||
| AK162595.1 | 5 | 1458 | ||
| BC010971.1 | 9 | 1774 | AAH10971.1 |
More Information
Set Membership: |
|

Genomic Map