Assay Details
Target Gene Details
Entrez Gene ID: | 19727 |
Gene Name: | regulatory factor X-associated ankyrin-containing protein |
Gene Aliases: |
Tvl1 |
Location: |
Chr.8:70130806-70139197 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rfxank | NM_001025589.1 | 3 | 514 | NP_001020760.1 |
| NM_011266.2 | 3 | 514 | NP_035396.1 | |
| AF094761.1 | 3 | 430 | AAC69884.1 | |
| AF123704.1 | 3 | 415 | AAD24798.1 | |
| AK052298.1 | 3 | 471 | BAC34924.1 | |
| AK087462.1 | 3 | 443 | ||
| AK146148.1 | 3 | 443 | BAE26933.1 | |
| BC010971.1 | 3 | 448 | AAH10971.1 | |
| CN526109.1 | 3 | 514 | ||
| L43164.1 | 3 | 415 | AAL77617.1 |
More Information
Set Membership: |
|

Genomic Map