Assay Details
Target Gene Details
Entrez Gene ID: | 59005 |
Gene Name: | trafficking protein particle complex 2-like |
Gene Aliases: |
1810017G16Rik, AB030200, Hspc176, Tca17 |
Location: |
Chr.8:122611626-122615591 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Trappc2l | NM_021502.2 | NP_067477.1 | ||
| AB030200.1 | BAA92763.1 | |||
| AK007526.1 | BAB25089.1 | |||
| BC028502.1 | AAH28502.1 | |||
| BY091059.1 |
More Information
Set Membership: |
|

Genomic Map