Assay Details
Target Gene Details
Entrez Gene ID: | 67439 |
Gene Name: | XPA binding protein 2 |
Gene Aliases: |
0610041O14Rik, AV025587 |
Location: |
Chr.8:3610087-3621334 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xab2 | NM_026156.2 | NP_080432.1 | ||
| AK002890.1 | BAB22435.1 | |||
| AK087952.1 | ||||
| AK132434.1 | ||||
| AK145925.1 | BAE26756.1 | |||
| AK201772.1 | ||||
| BC021341.1 | AAH21341.1 |
More Information
Set Membership: |
|

Genomic Map