Assay Details
Target Gene Details
Entrez Gene ID: | 76568 |
Gene Name: | intraflagellar transport 46 |
Gene Aliases: |
1500035H01Rik, AA408110, AI663846, R75047, c11orf2 |
Location: |
Chr.9:44772938-44792714 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ift46 | NM_023831.3 | 1 | 121 | NP_076320.2 |
| AJ249981.1 | 1 | 165 | CAB96545.1 | |
| AK005360.1 | 1 | 129 | BAB23974.1 | |
| AK032880.1 | 1 | 122 | BAC28067.1 | |
| BC080764.1 | 1 | 63 | AAH80764.1 |

Genomic Map